Merck
  • A validated LC-MS/MS method for quantitative analysis of curcumin in mouse plasma and brain tissue and its application in pharmacokinetic and brain distribution studies.
A validated LC-MS/MS method for quantitative analysis of curcumin in mouse plasma and brain tissue and its application in pharmacokinetic and brain distribution studies.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-30)
Prakash Ramalingam, Young Tag Ko
ABSTRACT
Curcumin is a well-known multitherapeutic agent widely employed in neurodegenerative disorders and cancer. A selective, fast, and sensitive method employing liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS) was developed and validated for the simultaneous determination of curcumin in mouse plasma and brain tissue, by using salbutamol as an internal standard. Triple quadrupole mass detection with multiple reaction monitoring (MRM) mode was used to monitor the ion transitions, m/z of 369>285 for curcumin, and m/z of 240>148 for salbutamol. The method was validated for recovery, accuracy, precision, linearity, and applicability. The lower limits of quantification (LLOQ) in both matrices were 2.5ng/mL. The inter-day and intra-day precisions and accuracy values were within the Food and Drug Administration (FDA) acceptance criteria, for both matrixes. The method was successfully applied in pharmacokinetics and brain distribution studies of curcumin after intravenous administration of free curcumin and curcumin-loaded solid lipid nanoparticles to mice. Furthermore, the application of this method along with serial blood sampling in mice has led to significant reduction in animal use and dosage and drastic improvement in speed, throughput, and quality of pharmacokinetic parameters.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Curcumin, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Salbutamol
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Salbutamol, VETRANAL®, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
Supelco
Curcumin, analytical standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Salbutamol, European Pharmacopoeia (EP) Reference Standard
Curcumin, primary reference standard
Supelco
Acetonitrile, analytical standard
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%