Merck
  • A fatal intoxication of 2,5-dimethoxy-4-chloroamphetamine: a case report.
A fatal intoxication of 2,5-dimethoxy-4-chloroamphetamine: a case report.
Journal of analytical toxicology (2014-09-14)
Rachel Y Barnett, Daniel D Baker, Nancy E Kelly, Calvin E McGuire, Timothy C Fassette, Jan M Gorniak
ABSTRACT
Designer drugs appear to be increasing in popularity because of the ease of obtaining these constituents, the lack of ability to identify the substance(s) in routine drug screening, the appeal of the drug(s) being 'safe' due to them being marketed as a 'legal high' and possibly due to stronger restrictions that are being placed on prescription drugs. As components of designer drugs are identified and regulated by the DEA, new constituents, or analogs, of these designer drugs are being manufactured to circumvent legislation. 2,5-Dimethoxy-4-chloroamphetamine (DOC) is a substituted alpha-methylated phenethylamine and acts as a selective serotonin receptor partial agonist. There is limited literature on this particular compound and no literature that attributes death to use of this drug alone. We present a case of a 37-year-old male found at home lying face down next to a book titled 'Psychedelic Chemistry' by Michael Valentine Smith and in the early stages of decomposition. The decedent was a known methamphetamine abuser. A peripheral blood sample collected at autopsy was sent to toxicology for routine analysis. Results yielded negative for the drugs of abuse classes on the enzyme-linked immunosorbent assay screen but was positive for DOC during routine GC-MS analysis. A urine sample collected at autopsy was subjected to a routine urine liquid/liquid analysis via GC-MS, and the specimen was positive for DOC. Quantification analyses showed DOC concentration levels to be 377 ng/mL in iliac blood; 3,193 ng/mL in urine; 3,143 ng/g in liver and 683 ng/g in brain. DOC was not detected in the gastric contents. Caffeine was the only other compound detected in blood and urine. Due to the lack of literature, we believe that this is the first case where death can be attributed to DOC alone.
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous