Merck
  • Decreased serum levels of free fatty acids are associated with breast cancer.
Decreased serum levels of free fatty acids are associated with breast cancer.
Clinica chimica acta; international journal of clinical chemistry (2014-07-13)
Yaping Zhang, Lina Song, Ning Liu, Chengyan He, Zhili Li
ABSTRACT
Changes in the levels of lipids are associated with breast cancer (BC). Disease-specific serum free fatty acids (FFAs) were quantified using chip-based direct-infusion nanoelectrospray ionization-Fourier transform ion cyclotron resonance mass spectrometry (CBDInanoESI-FTICR MS) in the negative ion mode. Multiple point internal standard calibration curves between the concentration ratios of fatty acids (i.e., C16:1, C18:3, C18:2, C18:1, C20:4, and C22:6) to internal standards (C17:1 for C16:1, C18:3, C18:2, and C18:1, C21:0 for C20:4 and C22:6) and their corresponding intensity ratios were established with a correlation coefficient of greater than 0.986. Data from 342 serum samples including 202 healthy controls and 140 BC patients indicate that serum concentrations of FFAs in patients with BC were significantly decreased compared with those in healthy controls. A panel of C16:1, C18:3, C18:2, C20:4, and C22:6 showed an excellent diagnostic ability to differentiate the patients with early stage BC from healthy controls, with the area under the receiver operating characteristics (ROC) curve of 0.953, a sensitivity of 83.3%, and a specificity of 87.1%. Our findings suggest that these FFAs may be a valuable biomarker panel for the early-stage detection of BC.
Sigma-Aldrich
Ultrapure Acetonitrile
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%