Merck
  • Environmental application of millimeter-scale sponge iron (s-Fe(0)) particles (II): the effect of surface copper.
Environmental application of millimeter-scale sponge iron (s-Fe(0)) particles (II): the effect of surface copper.
Journal of hazardous materials (2015-02-11)
Yongming Ju, Xiaowen Liu, Runlong Liu, Guohua Li, Xiaoyan Wang, Yanyan Yang, Dongyang Wei, Jiande Fang, Dionysios D Dionysiou
ABSTRACT
To enhance the catalytic reactivity of millimeter-scale particles of sponge iron (s-Fe(0)), Cu(2+) ions were deposited on the surface of s-Fe(0) using a simple direct reduction reaction, and the catalytic properties of the bimetallic system was tested for removal of rhodamine B (RhB) from an aqueous solution. The influence of Cu(0) loading, catalyst dosage, particle size, initial RhB concentration, and initial pH were investigated, and the recyclability of the catalyst was also assessed. The results demonstrate that the 3∼5 millimeter s-Fe(0) particles (s-Fe(0)(3∼5mm)) with 5wt% Cu loading gave the best results. The removal of RhB followed two-step, pseudo-first-order reaction kinetics. Cu(0)-s-Fe(0) showed excellent stability after five reuse cycles. Cu(0)-s-Fe(0) possesses great advantages compared to nanoscale zero-valent iron, iron power, and iron flakes as well as its bimetals. The surface Cu(0) apparently catalyzes the production of reactive hydrogen atoms for indirect reaction and generates Fe-Cu galvanic cells that enhance electron transfer for direct reaction. This bimetallic catalyst shows great potential for the pre-treatment of recalcitrant wastewaters. Additionally, some oxides containing iron element are selected to simulate the adsorption process. The results prove that the adsorption process of FeOOH, Fe2O3 and Fe3O4 played minor role for the removal of RhB.
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Supelco
Rhodamine B, analytical standard
Sigma-Aldrich
Rhodamine B, Dye content 90 %
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
Rhodamine B, suitable for fluorescence