Merck
  • An integrated quantitative and targeted proteomics reveals fitness mechanisms of Aeromonas hydrophila under oxytetracycline stress.
An integrated quantitative and targeted proteomics reveals fitness mechanisms of Aeromonas hydrophila under oxytetracycline stress.
Journal of proteome research (2015-01-27)
Xiangmin Lin, Ling Lin, Zujie Yao, Wanxin Li, Lina Sun, Danfeng Zhang, Ji Luo, Wenxiong Lin
ABSTRACT
To date, above ten thousand tons of antibiotics are used in aquaculture each year that lead to the deterioration of natural resources. However, knowledge is limited on the molecular biological behavior of common aquatic pathogens against antibiotics stress. In this study, proteomics profiles of Aeromonas hydrophila, which were exposed to different levels of oxytetracycline (OXY) stress, were displayed and compared using iTRAQ labeling and SWATH-MS based LC-MS/MS methods. A total 1383 proteins were identified by SWATH-MS method, and 2779 proteins were identified from iTRAQ labeling samples. There are 152 up-regulated and 52 down-regulated proteins overlapped in 5 μg/mL OXY stress and both 83 up- and down-regulated proteins overlapped in 10 μg/mL OXY stress in both methods, respectively. Results show that many protein synthesis and translation related proteins increased, while energy generation related proteins decreased in OXY stress. The varieties of selected proteins involved in both pathways were further validated by sMRM(HR), q-PCR, and enzyme activity assay. Furthermore, the concentrations of NAD+ and NADH were measured to verify the characteristic of energy generation process in OXY stress and OXY resistance strain. We demonstrate that the down-regulation of energy generation related metabolic pathways and up-regulation of translation may play an important role in antibiotics fitness or resistance of aquatic pathogens.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Proline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
3-Indoleacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
D-Glutamine, ≥98% (HPLC)
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%