Merck
  • Asymmetric trapping of zwitterionic intermediates by sulphur ylides in a palladium-catalysed decarboxylation-cycloaddition sequence.
Asymmetric trapping of zwitterionic intermediates by sulphur ylides in a palladium-catalysed decarboxylation-cycloaddition sequence.
Nature communications (2014-11-22)
Tian-Ren Li, Fen Tan, Liang-Qiu Lu, Yi Wei, Ya-Ni Wang, Yi-Yin Liu, Qing-Qing Yang, Jia-Rong Chen, De-Qing Shi, Wen-Jing Xiao
ABSTRACT
Through nearly 50 years of development, sulphur ylides have been established as versatile and powerful reagents for the construction of carbocycles and heterocycles. Despite advances, two important and yet elusive bottlenecks continue to inhibit the advancement of this chemistry: a limited number of reagents with polar groups to react with sulphur ylides, and the wide utilization of chiral auxiliaries or substrates to achieve asymmetric cycloaddition processes in the majority of known reports. Herein, we apply an asymmetric palladium catalysis strategy to the chemistry of sulphur ylides to address these two fundamental problems. We thus achieve an unprecedented decarboxylation-cycloaddition sequence of cyclic allylic esters with sulphur ylides through the enantioselective trapping of Pd-stabilized zwitterionic intermediates by the ylides. As a result, a series of biologically and synthetically important 3-vinyl indolines are rapidly assembled with a high reaction efficiency and stereoselectivity.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Propanol, analytical standard
Supelco
Hexane, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)