Merck
  • Mutations in PNPLA6 are linked to photoreceptor degeneration and various forms of childhood blindness.
Mutations in PNPLA6 are linked to photoreceptor degeneration and various forms of childhood blindness.
Nature communications (2015-01-13)
S Kmoch, J Majewski, V Ramamurthy, S Cao, S Fahiminiya, H Ren, I M MacDonald, I Lopez, V Sun, V Keser, A Khan, V Stránecký, H Hartmannová, A Přistoupilová, K Hodaňová, L Piherová, L Kuchař, A Baxová, R Chen, O G P Barsottini, A Pyle, H Griffin, M Splitt, J Sallum, J L Tolmie, J R Sampson, P Chinnery, E Banin, D Sharon, S Dutta, R Grebler, C Helfrich-Foerster, J L Pedroso, D Kretzschmar, M Cayouette, R K Koenekoop
ABSTRACT
Blindness due to retinal degeneration affects millions of people worldwide, but many disease-causing mutations remain unknown. PNPLA6 encodes the patatin-like phospholipase domain containing protein 6, also known as neuropathy target esterase (NTE), which is the target of toxic organophosphates that induce human paralysis due to severe axonopathy of large neurons. Mutations in PNPLA6 also cause human spastic paraplegia characterized by motor neuron degeneration. Here we identify PNPLA6 mutations in childhood blindness in seven families with retinal degeneration, including Leber congenital amaurosis and Oliver McFarlane syndrome. PNPLA6 localizes mostly at the inner segment plasma membrane in photoreceptors and mutations in Drosophila PNPLA6 lead to photoreceptor cell death. We also report that lysophosphatidylcholine and lysophosphatidic acid levels are elevated in mutant Drosophila. These findings show a role for PNPLA6 in photoreceptor survival and identify phospholipid metabolism as a potential therapeutic target for some forms of blindness.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
DAPI, for nucleic acid staining
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard