Merck
  • Isolation & identification of bacteria for the treatment of brown crab (Cancer pagurus) waste to produce chitinous material.
Isolation & identification of bacteria for the treatment of brown crab (Cancer pagurus) waste to produce chitinous material.
Journal of applied microbiology (2015-02-04)
C Harkin, W M Brück, C Lynch
ABSTRACT
To isolate bacteria from soil for microbial pretreatment of brown crab (Cancer pagurus) shell waste and the production of chitin. Isolates were screened for protease enzymes and acid production in order to facilitate the removal of protein and calcium carbonate fractions from brown crab shell to yield a chitinous material. Selected isolates were applied in various combinations in successive, two-step fermentations with brown crab shell waste. These isolates were identified as: Exiguobacterium spp. (GenBank accession number: KP050496), Bacillus cereus (GenBank accession number: KP050499), B. subtilis (GenBank accession number: KP050498), Bacillus licheniformis (GenBank accession number: KP050497), Pseudomonas migulae (GenBank accession number: KP050501), Pseudomonas spp. (GenBank accession number: KP050500), Pseudomonas spp. (GenBank accession number: KP050502), Arthrobacter luteolus (GenBank accession number: KP050503), Lactobacillus spp. (GenBank accession number: KP072000) and Enterococcus spp. (GenBank accession number: KP071999). Successive two-step fermentations with isolates in certain combinations resulted in a demineralization of >94% and the extraction of a crude chitin fraction from brown crab processing waste. The highest demineralization, 98·9% was achieved when isolates identified as B. cereus and Pseudomonas spp. were used in combination. The transfer of fermentations to a larger scale requires further research for optimization. The successful application of these isolates in successive two-step fermentation of brown crab shell waste to extract chitin means with further research into optimization and scale up, this chitin extraction process may be applied on an industrial scale and provide further commercial value from brown crab shell waste.
Sigma-Aldrich
DL-Malic acid, 99%
Pricing and availability is not currently available.
Millipore
Bifido Selective Supplement B, suitable for microbiology
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Pricing and availability is not currently available.
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Pricing and availability is not currently available.
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Pricing and availability is not currently available.
Sigma-Aldrich
Malic acid, meets USP/NF testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Malic acid, ≥98% (capillary GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Pricing and availability is not currently available.
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Pricing and availability is not currently available.