Merck
  • Characterization of surface confined ionic liquid stationary phases: impact of cation revisited.
Characterization of surface confined ionic liquid stationary phases: impact of cation revisited.
Journal of chromatography. A (2014-09-15)
Bradley J VanMiddlesworth, Apryll M Stalcup
ABSTRACT
Modification of the Linear Solvation Energy Relationship (LSER) equation to account for ionic interactions in the retention of ionizable compounds has enabled the elucidation in the effect of the imidazolium cation identity on retention. Three Surface Confined Ionic Liquid stationary phases were synthesized from an octylbromide phase on silica: 1-octyl-3-methylimidazolium bromide (MIM), 1-octyl-3-butylimidazolium bromide (BIM), and 1-octyl-3-benzylimidazolium bromide (BzIM). These phases were probed via a 35 analyte probe set, including 6 phenolic acids, 5 anilinic bases, and 2 pyridinic bases, and the resulting column parameters compared with previously reported interactions of ionic liquids or Surface Confined Ionic Liquids. The correlation between experimental and calculated retention for the conventional, 6-parameter LSER equation was very poor: r(2)=0.64 (MIM), 0.60 (BIM), and 0.62 (BzIM). By accounting for the ionic interactions between stationary phase and analytes, linearity for the modified, 8 parameter LSER equation was significantly improved to r(2)=0.997 (MIM), 0.996 (BIM), and 0.997 (BzIM). The primary difference between cation identities is within the retention of acids where BIM>BzIM>MIM. We conjecture that the accessibility of bulky, acidic analytes to the on-top interaction of the imidazolium ring is the major contributor to increased anion retention.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Trichlorosilane, 99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Octene, 98%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Imidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)