Merck
  • Promotion of mesenchymal-to-epithelial transition by Rac1 inhibition with small molecules accelerates hepatic differentiation of mesenchymal stromal cells.
Promotion of mesenchymal-to-epithelial transition by Rac1 inhibition with small molecules accelerates hepatic differentiation of mesenchymal stromal cells.
Tissue engineering. Part A (2015-01-28)
Nan-Yuan Teng, Yi-Shiuan Liu, Hao-Hsiang Wu, Yu-An Liu, Jennifer H Ho, Oscar Kuang-Sheng Lee
ABSTRACT
In vitro differentiation of stem cells into specific cell lineages provides a stable cell supply for cell therapy and tissue engineering. Therefore, understanding the mechanisms underlying such differentiation processes is critical for generating committed lineage-specific cell progenies effectively. We previously developed a two-step protocol to differentiate mesenchymal stromal cells (MSCs) into hepatocyte-like cells. Since hepatic differentiation involves mesenchymal-epithelial transition (MET), we hypothesize that promoting MET could further accelerate the differentiation process. Ras-related C3 botulinum toxin substrate 1 (Rac1) is involved in actin polymerization and its role in MET was investigated in the study. Our results showed that inhibition of Rac1 activation by Rac1-specific inhibitor, NSC23766, led to cells favoring epithelial morphology and being more packed during hepatic differentiation. In addition, Rac1 inhibition accelerated the upregulation of hepatic marker genes accompanied by more mature hepatic functions. Taken together, promotion of MET by inhibiting Rac1 accelerates the hepatic differentiation of MSCs. Our findings open a new prospect of directing the commitment of MSCs by manipulating cell morphology and cytoskeleton arrangement through small molecules. The results provide further insight into scaffold design for rapid production of MSC-differentiated hepatocytes.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Millipore
Urea solution, suitable for microbiology, 40% in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Periodic acid, 99.999% trace metals basis
Sigma-Aldrich
Periodic acid, suitable for electrophoresis, ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source