Merck
  • Porphyromonas gingivalis gingipains selectively reduce CD14 expression, leading to macrophage hyporesponsiveness to bacterial infection.
Porphyromonas gingivalis gingipains selectively reduce CD14 expression, leading to macrophage hyporesponsiveness to bacterial infection.
Journal of innate immunity (2014-09-18)
Asaf Wilensky, Rinat Tzach-Nahman, Jan Potempa, Lior Shapira, Gabriel Nussbaum
ABSTRACT
Cysteine proteases (gingipains) from Porphyromonas gingivalis are key virulence factors in chronic periodontitis. Innate immune receptors CD14, Toll-like receptor (TLR) 2 and TLR4 are important in P. gingivalis recognition. We examined the ability of gingipains to cleave CD14, TLR2 and TLR4, and the consequences for the cellular response to bacterial challenge. Macrophages were exposed to Arg (RgpA and RgpB)- and Lys (Kgp)-gingipains, and residual expression of TLR2, TLR4 and CD14 was determined by flow cytometry. The cellular response to live bacteria following exposure to purified gingipains was evaluated by TNFα production and bacterial phagocytosis. RgpA and Kgp decreased CD14 detection in a concentration (p = 0.0000002)- and time (p = 0.03)-dependent manner, whereas RgpB had no significant effect. TLR2 and TLR4 expression were unaffected. Reduction in CD14 expression was more efficient with Lys-gingipain than with Arg-gingipain. A reduced CD14 surface level correlated with decreased TNFα secretion and bacterial phagocytosis following challenge with live P. gingivalis, but the response to heat-killed bacteria was unaffected. Therefore, gingipains reduce CD14 expression without affecting expression of the bacterial-sensing TLRs. Reduced CD14 expression depends on the gingipain hemagglutinin/adhesion site and results in macrophage hyporesponsiveness to bacterial challenge. Further studies are needed to determine if reduced CD14 expression is linked to periodontitis induced by P. gingivalis.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
DL-Cysteine, technical grade