Merck
  • Design, synthesis and evaluation of a gonadotropin releasing hormone-based subunit vaccine in rams (Ovis aries).
Design, synthesis and evaluation of a gonadotropin releasing hormone-based subunit vaccine in rams (Ovis aries).
Vaccine (2015-02-14)
Daryn Goodwin, Pegah Varamini, Pavla Simerska, Michael J D'Occhio, Istvan Toth
ABSTRACT
Immunocastration using gonadotropin-releasing hormone (GnRH)-based vaccines has been investigated in rams to reduce aggressive and sexual behaviour and to control meat quality. Despite considerable efforts, a practical GnRH vaccine has yet to be developed for rams. In the present study, a A GnRH-lipopeptide vaccine (GnRH-LP) including two copies of GnRH, 2-amino-d,l-hexadecanoic acid (C16), and a unique T helper epitope, was examined in rams. Rams received a primary and secondary vaccination of GnRH-LP without additional adjuvant (Group 1) or with the adjuvant AdjuVac™ (Group 2). In both Group 1 and 2 anti-GnRH antibody titres increased after secondary vaccination, however, the antibody titres were higher (p<0.01) for rams in Group 2. The latter rams showed a marked decrease in testicular size. The marked and sustained reduction in testicular size in rams treated with GnRH-LP+AdjuVac™ provides the basis for an effective immunocastration vaccine in rams.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, for ultratrace analysis, ACS reagent
Supelco
N,N-Dimethylformamide, analytical standard
Supelco
Methanol, analytical standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
N-Ethyldiisopropylamine, ≥98.0%
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, peroxidase substrate
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 2 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 30 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 1 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 20 mg substrate per tablet
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 4 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 15 mg substrate per tablet
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture