Merck
  • Immunosuppressive activity of alpinetin on activation and cytokines secretion of murine T lymphocytes.
Immunosuppressive activity of alpinetin on activation and cytokines secretion of murine T lymphocytes.
Immunopharmacology and immunotoxicology (2014-06-27)
Shuang Guan, Baochen Fang, Bocui Song, Ying Xiong, Jing Lu
ABSTRACT
Abstract Alpinetin, a flavonoid compound extracted from the seeds of Alpinia katsumadai Hayata, has been known to possess antibacterial, anti-inflammatory and other important therapeutic activities. In the current study, we investigated alpinetin for its immunosuppressive effect on activation and cytokines secretion of murine T lymphocytes. The data showed that alpinetin markedly suppressed ConA-induced murine splenocyte proliferation, Th1/Th2 cytokines production, CD4(+) T-cell populations and ratio of CD4(+)/CD8(+). This inspired us to further study the effects of alpinetin in vivo. The results showed that administration of alpinetin suppressed T-cell-mediated delayed-type hypersensitivity reaction in mice. In addition, we studied signal transduction pathways about T-cell activation on puried murine T lymphocytes by Western-blot assay. The data revealed that alpinetin could shock the activation of NF-κB, NFAT2 signal transduction pathways. These observations indicated that alpinetin have potential effects in downregulating the immune system and might be developed as a useful immunosuppressive agent in treating undesired immune responses.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Human IL-2 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Bovine IL2 / Interleukin-2 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Mouse IL-2 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat IL-2 ELISA Kit, for serum, plasma, cell culture supernatant