Merck
Anti-HCV activity of the Chinese medicinal fungus Cordyceps militaris.
Biochemical and biophysical research communications (2014-04-15)
Youki Ueda, Kyoko Mori, Shinya Satoh, Hiromichi Dansako, Masanori Ikeda, Nobuyuki Kato
ABSTRACT
Persistent hepatitis C virus (HCV) infection causes chronic liver diseases and is a global health problem. Although the sustained virologic response rate in the treatment of genotype 1 using new triple therapy (pegylated-interferon, ribavirin, and telaprevir/boceprevir) has been improved by more than 70%, several severe side effects such as skin rash/ageusia and advanced anemia have become a problem. Under these circumstances, a new type of anti-HCV oral drug with few side effects is needed. Our recently developed HCV drug assay systems, including the HuH-7 cell line-derived OR6 and AH1R, and the Li23 cell line-derived ORL8 and ORL11, allow genome-length HCV RNAs (several strains of genotype 1b) encoding renilla luciferase to replicate efficiently. Using these systems as anti-HCV candidates, we have identified numerous existing medicines that can be used against HCV with few side effects, such as statins and teprenon. To obtain additional anti-HCV candidates, we evaluated a number of oral health supplements, and found that the capsule but not the liquid form of Cordyceps militaris (CM) (Ascomycotinanorth, North Chinese caterpillar fungus), which is used as a Chinese herbal medicine, exhibited moderate anti-HCV activity. In combination with interferon-α or ribavirin, CM exhibited an additive inhibitory effect. Among the main components of CM, cordycepin, but not ergosterol, contributed to the anti-HCV activity of CM. In consideration of all these results, we suggest that CM would be useful as an oral anti-HCV agent in combination with interferon-α and/or ribavirin.
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Ciclosporin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ergosterol, ≥75%
Sigma-Aldrich
(±)-CPP, solid
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
α-Tocopherol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
Cordycepin, from Cordyceps militaris
Supelco
Ergosterol, Pharmaceutical Secondary Standard; Certified Reference Material
Ergosterol, European Pharmacopoeia (EP) Reference Standard
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
USP
Alpha Tocopherol, United States Pharmacopeia (USP) Reference Standard