Merck
  • Triclosan exhibits a tendency to accumulate in the epididymis and shows sperm toxicity in male Sprague-Dawley rats.
Triclosan exhibits a tendency to accumulate in the epididymis and shows sperm toxicity in male Sprague-Dawley rats.
Environmental toxicology (2013-08-10)
Zhou Lan, Tae Hyung Kim, Kai Shun Bi, Xiao Hui Chen, Hyung Sik Kim
ABSTRACT
Triclosan (TCS) is considered a potent endocrine disruptor that causes reproductive toxicity in non-mammals, but it is still unclear exactly whether TCS has adverse effects on the sperm or reproductive organs in mammals. In this study, we aimed to evaluate the distribution status of TCS in male reproductive organs of rats, and seek the correlation with the TCS-induced sperm toxicity or reproductive organ damage. Male rats were intragastrically administered with TCS at a dose of 50 mg/kg, the kinetics of TCS in the plasma and reproductive organs were investigated. TCS in testes and prostates both showed a lower-level distribution compared to that in the plasma, which indicates it has no tendency to accumulate in those organs. However, TCS in the epididymides showed a longer elimination half-life (t1/2 z), a longer the mean retention time (MRT), and a lower clearance (CLZ /F) compared with those in the plasma. Besides, the ratios of mean area under the concentration-time curve (AUC)(0-96 h(epididymides/plasma)) and AUC(0-∞(epididymides/plasma)) were 1.13 and 1.51, respectively. These kinetic parameters suggest TCS has an accumulation tendency in the epididymides. Based on this, we investigated the TCS-induced sperm toxicity and histopathological changes of reproductive organs in rats. TCS was given intragastrically at doses of 10, 50, and 200 mg/kg for 8 weeks. Rats treated with the high dose (200 mg/kg) of TCS showed a significant decrease in daily sperm production (DSP), changes in sperm morphology and epididymal histopathology. Considering the histopathological change in the epididymides, TCS may induce the epididymal damage due to the epididymal accumulation of that.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Eosin Y disodium salt, ≥85 % by HPLC
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Eosin Y disodium salt, certified by the BSC
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Triclosan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
USP
Triclosan, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%