Merck
  • Enhancing neuronal growth from human endometrial stem cells derived neuron-like cells in three-dimensional fibrin gel for nerve tissue engineering.
Enhancing neuronal growth from human endometrial stem cells derived neuron-like cells in three-dimensional fibrin gel for nerve tissue engineering.
Journal of biomedical materials research. Part A (2013-08-22)
Mona Navaei-Nigjeh, Ghasem Amoabedini, Abbas Noroozi, Mahmoud Azami, Mohammad N Asmani, Somayeh Ebrahimi-Barough, Hooshang Saberi, Armin Ai, Jafar Ai
ABSTRACT
Nerve tissue engineering (NTE) is one of the most promising methods to restore central nerve systems in human health care. Three-dimensional (3D) distribution and growth of cells within the porous scaffold composed of nanofibers are of clinical significance for NTE. In this study, an attempt was made to develop and characterize the use of fibrin gel and human endometrial stem cells (hEnSCs)-derived neuron-like cells simultaneously to support cell behavior especially neuron outgrowth. The structural and mechanical characteristics of fibrin gel scaffold were examined with SEM and rheometer. Also, hEnSCs-derived neuron-like cells were cultured in fibrin gel and were subsequently analyzed with immunofluorescent staining against neuronal markers. In parallel, the survival and growth rates of the cells were determined by MTT assay and neurite extension. At the end, cell-matrix interactions were investigated with SEM and TEM micrographs. Mechanical properties of fabricated scaffold were studied and results indicated appropriate choice of material, SEM and TEM showed excellent integration of cells with nanofibers regarding the relation between cells and fibrin gel. Immunofluorescent staining of fibrin gel after 6 days of cell seeding and culture demonstrated well expanded and incorporated network of neurons. In addition, viability, proliferation, and neuronal growth of seeded cells were analyzed at days 1, 3, and 6. Comparing those results with 2D culture of seeded cells showed positive effect of 3D culture. Taken together, the results suggest that fibrin can provide a suitable, three-dimensional scaffold for neuronal survival and outgrowth for regeneration of the central nervous system.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glutaraldehyde solution, 25 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC