Merck
Early netrin-1 expression impairs central nervous system remyelination.
Annals of neurology (2014-06-20)
Vanja Tepavčević, Christophe Kerninon, Marie Stéphane Aigrot, Elodie Meppiel, Sabah Mozafari, Raphaelle Arnould-Laurent, Philippe Ravassard, Timothy E Kennedy, Brahim Nait-Oumesmar, Catherine Lubetzki
ABSTRACT
Chronically demyelinated multiple sclerosis (MS) lesions are frequently characterized by scarce undifferentiated oligodendrocyte progenitor cells (OPCs), suggesting the exhaustion of a local OPC pool followed by failure of recruitment and differentiation. Stimulating prompt OPC recruitment following demyelination could improve myelin repair by providing sufficient numbers of remyelinating cells during the repair-permissive period. Understanding mechanisms that determine this process may have important therapeutic implications. We therefore investigated the role of the guidance molecule netrin-1 in OPC recruitment and central nervous system (CNS) remyelination. Netrin-1 expression was analyzed immunohistochemically in different types of MS lesions and in the murine lysolecithin model of demyelination. The influence of netrin-1 on CNS remyelination was examined using gain and loss of function experiments. We show that in MS lesions, astrocytes upregulate netrin-1 expression early during demyelination and netrin-1 receptors are expressed by OPCs. In contrast, in the efficiently repairing lysolecithin model of demyelination (astrocyte-free), netrin-1 expression is absent during early phases and detected concomitant with completion of OPC recruitment. In vitro migration assays demonstrated that netrin-1 is a chemorepellent for migrating adult OPCs. In mouse lesions, antibody-mediated disruption of netrin-1 function at the peak phase of recruitment increased OPC numbers. Conversely, lentiviral-mediated induction of netrin-1 expression prior to OPC recruitment reduced the number of cells recruited and impaired remyelination. Our findings support the conclusion that netrin-1 expression within demyelinating MS plaques blocks OPC recruitment, which with repeated demyelinating episodes contributes to permanent remyelination failure.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, 25 wt. % in H2O
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-OLIG1 antibody produced in mouse, clone 2B11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse
Sigma-Aldrich
Anti-Olig2 Antibody, Chemicon®, from rabbit