Merck
  • Amide-type local anesthetics action on the sarcoplasmic reticulum Ca-ATPase from fast-twitch skeletal muscle.
Amide-type local anesthetics action on the sarcoplasmic reticulum Ca-ATPase from fast-twitch skeletal muscle.
Naunyn-Schmiedeberg's archives of pharmacology (2014-06-21)
D E Di Croce, P W Trinks, C de La Cal, G A Sánchez, D Takara
ABSTRACT
Myotoxic effects related to intracellular Ca(2+) disturbances have been reported for local anesthetics. Such effects might derive from Ca-ATPase dysfunction. The aim of this work was to describe the effect of lidocaine and bupivacaine on the sarcoplasmic reticulum (SR) Ca-ATPase from fast-twitch skeletal muscle and to identify the affected steps of the enzyme's cycle. SR sealed vesicles were isolated from rabbit fast-twitch muscles by ultracentrifugation. The effect of the anesthetics on Ca-ATPase activity was assessed with a colorimetric method and Ca(2+) binding, uptake, phosphorylation of the enzyme by ATP, Ca(2+) dissociation kinetics and phosphoenzyme formation and decomposition levels were tested with radioisotopic methods. Lidocaine and bupivacaine inhibited Ca-ATPase activity with half-maximal inhibitory concentrations (Ki) of 25.3 and 31.4 mM, respectively, and the steady-state Ca(2+) transport ability with Ki values of 33.6 and 46.5 mM, decreasing the maximal transport rate without modification of the Ca(2+) or ATP affinity for the enzyme. This is consistent with an absence of competition for the transport and catalytic sites. The anesthetics did not inhibit Ca(2+) binding but inhibited the phosphorylation partial reactions. Ca(2+) dissociation kinetics was not affected, but the phosphoenzyme levels were decreased, and the decomposition rate of the phosphoenzyme became faster in the presence of the anesthetics. It is concluded that lidocaine and bupivacaine at concentrations available in pharmaceutical formulations for clinical medical and dental uses inhibit the SR Ca-ATPase through inhibition of key phosphorylation steps of the enzymatic cycle.
Product Description
SAFC
MOPS
Sigma-Aldrich
Acetamide, ≥99.0% (GC)
Sigma-Aldrich
MOPS, BioUltra, Molecular Biology, ≥99.5% (titration)
Sigma-Aldrich
Calcium Ionophore A23187 mixed calcium magnesium salt, Approximate 1:15 molar ratio Ca:Mg.
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Acetamide, ~99% (GC)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Acetamide, sublimed, 99%
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O