Merck
  • GHSR DNA hypermethylation is a common epigenetic alteration of high diagnostic value in a broad spectrum of cancers.
GHSR DNA hypermethylation is a common epigenetic alteration of high diagnostic value in a broad spectrum of cancers.
Oncotarget (2015-01-06)
Evgeny A Moskalev, Pouria Jandaghi, Mahdi Fallah, Mehdi Manoochehri, Sandeep K Botla, Oleg V Kolychev, Evgeny A Nikitin, Vladymyr V Bubnov, M von Knebel Doeberitz, Oliver Strobel, Thilo Hackert, Markus W Büchler, Nathalia Giese, Andrea Bauer, Thomas Muley, Arne Warth, Peter Schirmacher, Florian Haller, Jörg D Hoheisel, Yasser Riazalhosseini
ABSTRACT
Identification of a single molecular trait that is determinant of common malignancies may serve as a powerful diagnostic supplement to cancer type-specific markers. Here, we report a DNA methylation mark that is characteristic of seven studied malignancies, namely cancers of lung, breast, prostate, pancreas, colorectum, glioblastoma and B cell chronic lymphocytic leukaemia (CLL) (n = 137). This mark was defined by substantial hypermethylation at the promoter and first exon of growth hormone secretagouge receptor (GHSR) through bisulfite pyrosequencing. The degree of aberrant methylation was capable of accurate discrimination between cancer and control samples. The highest sensitivity and specificity of cancer detection was achieved for cancers of pancreas, lung, breast and CLL yielding the area under the curve (AUC) values of 1.0000, 0.9952, 0.9800 and 0.9400, respectively. Narrowing to a single CpG site within the gene's promoter or four consecutive CpG units of the highest methylation levels within the first exon improved the detection power. GHSR hypermethylation was detected already at the early stage tumors. The accurate performance of this marker was further replicated in an independent set of pancreatic cancer and control samples (n = 78). These findings support the candidature of GHSR methylation as a highly accurate pan-cancer marker.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%