Merck
  • Infection by Toxoplasma gondii, a severe parasite in neonates and AIDS patients, causes impaired anion secretion in airway epithelia.
Infection by Toxoplasma gondii, a severe parasite in neonates and AIDS patients, causes impaired anion secretion in airway epithelia.
Proceedings of the National Academy of Sciences of the United States of America (2015-04-02)
Hong-Mei Guo, Jiang-Mei Gao, Yu-Li Luo, Yan-Zi Wen, Yi-Lin Zhang, Geoff Hide, Wen-Liang Zhou, Francisco J Ayala, Zhao-Rong Lun
ABSTRACT
The airway epithelia initiate and modulate the inflammatory responses to various pathogens. The cystic fibrosis transmembrane conductance regulator-mediated Cl(-) secretion system plays a key role in mucociliary clearance of inhaled pathogens. We have explored the effects of Toxoplasma gondii, an opportunistic intracellular protozoan parasite, on Cl(-) secretion of the mouse tracheal epithelia. In this study, ATP-induced Cl(-) secretion indicated the presence of a biphasic short-circuit current (Isc) response, which was mediated by a Ca(2+)-activated Cl(-) channel (CaCC) and the cystic fibrosis transmembrane conductance regulator. However, the ATP-evoked Cl(-) secretion in T. gondii-infected mouse tracheal epithelia and the elevation of [Ca(2+)]i in T. gondii-infected human airway epithelial cells were suppressed. Quantitative reverse transcription-PCR revealed that the mRNA expression level of the P2Y2 receptor (P2Y2-R) increased significantly in T. gondii-infected mouse tracheal cells. This revealed the influence that pathological changes in P2Y2-R had on the downstream signal, suggesting that P2Y2-R was involved in the mechanism underlying T. gondii infection in airways. These results link T. gondii infection as well as other pathogen infections to Cl(-) secretion, via P2Y2-R, which may provide new insights for the treatment of pneumonia caused by pathogens including T. gondii.
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Diphenylcyclopropenone, 98%
Sigma-Aldrich
U-73122 hydrate, powder
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of 30 mg
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of ~1 mg ATP
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade II, ≥97% (HPLC), crystalline, from microbial
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, microbial, BioReagent, suitable for cell culture, ≥99% (HPLC)
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial
Sigma-Aldrich
Suramin sodium salt, ≥98% (HPLC)
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%