Merck
  • Proteogenomics analysis reveals specific genomic orientations of distal regulatory regions composed by non-canonical histone variants.
Proteogenomics analysis reveals specific genomic orientations of distal regulatory regions composed by non-canonical histone variants.
Epigenetics & chromatin (2015-04-17)
Kyoung-Jae Won, Inchan Choi, Gary LeRoy, Barry M Zee, Simone Sidoli, Michelle Gonzales-Cope, Benjamin A Garcia
ABSTRACT
Histone variants play further important roles in DNA packaging and controlling gene expression. However, our understanding about their composition and their functions is limited. Integrating proteomic and genomic approaches, we performed a comprehensive analysis of the epigenetic landscapes containing the four histone variants H3.1, H3.3, H2A.Z, and macroH2A. These histones were FLAG-tagged in HeLa cells and purified using chromatin immunoprecipitation (ChIP). By adopting ChIP followed by mass spectrometry (ChIP-MS), we quantified histone post-translational modifications (PTMs) and histone variant nucleosomal ratios in highly purified mononucleosomes. Subsequent ChIP followed by next-generation sequencing (ChIP-seq) was used to map the genome-wide localization of the analyzed histone variants and define their chromatin domains. Finally, we included in our study large datasets contained in the ENCODE database. We newly identified a group of regulatory regions enriched in H3.1 and the histone variant associated with repressive marks macroH2A. Systematic analysis identified both symmetric and asymmetric patterns of histone variant occupancies at intergenic regulatory regions. Strikingly, these directional patterns were associated with RNA polymerase II (PolII). These asymmetric patterns correlated with the enhancer activities measured using global run-on sequencing (GRO-seq) data. Our studies show that H2A.Z and H3.3 delineate the orientation of transcription at enhancers as observed at promoters. We also showed that enhancers with skewed histone variant patterns well facilitate enhancer activity. Collectively, our study indicates that histone variants are deposited at regulatory regions to assist gene regulation.
Sigma-Aldrich
Propionic anhydride, ≥99%
Sigma-Aldrich
Propionic anhydride, purum, ≥96.0% (NT)
Sigma-Aldrich
Propionic anhydride, 97%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis