Merck
  • A growth-promoting signaling component cyclin D1 in neural stem cells has antiastrogliogenic function to execute self-renewal.
A growth-promoting signaling component cyclin D1 in neural stem cells has antiastrogliogenic function to execute self-renewal.
Stem cells (Dayton, Ohio) (2013-12-05)
Norihisa Bizen, Toshihiro Inoue, Takeshi Shimizu, Kouichi Tabu, Tetsushi Kagawa, Tetsuya Taga
ABSTRACT
Self-renewing proliferation of neural stem cells (NSCs) is intimately linked to the inhibition of neuronal and glial differentiation, however, their molecular linkage has been poorly understood. We have proposed a model previously explaining partly this linkage, in which fibroblast growth factor 2 (FGF2) and Wnt signals cooperate to promote NSC self-renewal via β-catenin accumulation, which leads to the promotion of proliferation by lymphoid enhancer factor (LEF)/T-cell factor (TCF)-mediated cyclin D1 expression and at the same time to the inhibition of neuronal differentiation by β-catenin-mediated potentiation of Notch signaling. To fully understand the mechanisms underlying NSC self-renewal, it needs to be clarified how these growth factor signals inhibit glial differentiation as well. Here, we demonstrate that cyclin D1, a NSC growth promoting signaling component and also a common component of FGF2 and Wnt signaling pathways, inhibits astroglial differentiation of NSCs. Interestingly, this effect of cyclin D1 is mediated even though its cell cycle progression activity is blocked. Forced downregulation of cyclin D1 enhances astrogliogenesis of NSCs in culture and in vivo. We further demonstrate that cyclin D1 binds to STAT3, a transcription factor downstream of astrogliogenic cytokines, and suppresses its transcriptional activity on the glial fibrillary acidic protein (Gfap) gene. Taken together with our previous finding, we provide a novel molecular mechanism for NSC self-renewal in which growth promoting signaling components activated by FGF2 and Wnts inhibit neuronal and glial differentiation.
Product Description
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
MISSION® esiRNA, targeting human CCND1
Sigma-Aldrich
Leukemia Inhibitory Factor from mouse, LIF, recombinant, expressed in E. coli, 10 μg/ml, buffered aqueous solution, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ccnd1