Merck
  • Punarnavine, an alkaloid isolated from ethanolic extract of Boerhaavia diffusa Linn. reverses depression-like behaviour in mice subjected to chronic unpredictable mild stress.
Punarnavine, an alkaloid isolated from ethanolic extract of Boerhaavia diffusa Linn. reverses depression-like behaviour in mice subjected to chronic unpredictable mild stress.
Indian journal of experimental biology (2014-08-22)
Dinesh Dhingra, Rekha Valecha
ABSTRACT
Punarnavine (20 and 40 mg/kg) and fluoxetine (20 mg/kg) per se administered orally for 14 successive days significantly decreased immobility periods of both unstressed and stressed mice in forced swim test. These drugs also significantly decreased sucrose preference in both stressed and unstressed mice as compared to their respective controls, indicating significant antidepressant-like activity. The drugs did not show any significant effect on locomotor activity of mice. The alkaloid also significantly decreased monoamine oxidase (MAO-A) activity, malondialdehyde levels in both unstressed and stressed mice; and significantly reversed the stress-induced decrease in reduced glutathione and catalase activity. It also significantly attenuated the stress-induced increase in plasma nitrite and corticosterone levels. Thus, punarnavine showed antidepressant-like activity in unstressed and stressed mice probably through inhibition of brain MAO-A activity, decrease in plasma nitrite levels and due to its antioxidant activity. In addition, punarnavine also showed antidepressant-like activity in stressed mice possibly through decrease in plasma corticosterone levels.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Fluoxetine hydrochloride, solid
USP
Fluoxetine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Fluoxetine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Fluoxetine hydrochloride, VETRANAL®, analytical standard