Merck
  • Photocatalytic degradation of carbamazepine in wastewater by using a new class of whey-stabilized nanocrystalline TiO2 and ZnO.
Photocatalytic degradation of carbamazepine in wastewater by using a new class of whey-stabilized nanocrystalline TiO2 and ZnO.
The Science of the total environment (2014-04-15)
D P Mohapatra, S K Brar, R Daghrir, R D Tyagi, P Picard, R Y Surampalli, P Drogui
ABSTRACT
Nanoscale photocatalysts have attracted much attention due to their high surface area to volume ratios. However, due to extremely high reactivity, TiO2 and ZnO nanoparticles prepared using different methods tend to either react with surrounding media or agglomerate, resulting in the formation of much larger flocs and significant loss in reactivity. This work investigates the photocatalytic degradation of carbamazepine (CBZ), a persistent pharmaceutical compound from wastewater (WW) using TiO2 and ZnO nanoparticles prepared in the presence of a water-soluble whey powder as stabilizer. The TiO2 and ZnO nanoparticles prepared in the presence of whey stabilizer displayed much less agglomeration and greater degradation power than those prepared without a stabilizer. Higher photocatalytic degradation of carbamazepine was observed (100%) by using whey stabilized TiO2 nanoparticles with 55 min irradiation time as compared to ZnO nanoparticles (92%). The higher degradation of CBZ in wastewater by using TiO2 nanoparticles as compared to ZnO nanoparticles was due to formation of higher photo-generated holes with high oxidizing power of TiO2. The photocatalytic capacity of ZnO anticipated as similar to that of TiO2 as it has the same band gap energy (3.2 eV) as TiO2. However, in the case of ZnO, photocorrosion frequently occurs with the illumination of UV light and this phenomenon is considered as one of the main reasons for the decrease of ZnO photocatalytic activity in aqueous solutions. Further, the estrogenic activity of photocatalyzed WW sample with CBZ and its by-products was carried out by yeast estrogen screen (YES) assay method. Based upon the YES test results, none of the samples showed estrogenic activity.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carbamazepine, analytical standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Carbamazepine, meets USP testing specifications