Merck
  • Insulin-like growth factor binding protein 4 enhances cardiomyocytes induction in murine-induced pluripotent stem cells.
Insulin-like growth factor binding protein 4 enhances cardiomyocytes induction in murine-induced pluripotent stem cells.
Journal of cellular biochemistry (2014-03-13)
Yuanyuan Xue, Yuan Yan, Hui Gong, Bo Fang, Yin Zhou, Zhiwen Ding, Peipei Yin, Guoping Zhang, Yong Ye, Chunjie Yang, Junbo Ge, Yunzeng Zou
ABSTRACT
Insulin-like growth factor binding protein 4 (IGFBP4) has been reported to play critical role in cardiomyocytes differentiation of embryonic stem cells (ESCs). But whether it promotes cardiomyocytes induction of iPSCs is unclear. In the present study, we aim to explore the role of IGFBP4 in the cardiogenesis of mouse iPSCs. We observed that IGFBP4 treatment at late stage during differentiation process of mouse iPSCs greatly enhanced the beating frequency of embryoid bodies (EBs). The expressions of Nkx2.5 (cardiac-specific transcription factor), α-MHC, α-actinin, and Troponin I (cardiac-specific protein) were significantly enhanced by IGFBP4 treatment. Immunostaining analysis showed that α-MHC, TNNT2 and connexin 43, typical cardiac markers, were obviously expressed in isolated cardiomyocytes from iPSCs with or without IGFBP4 treatment. Further study revealed that IGFBP4 had little effect on the apoptosis of EBs, but it significantly promoted the proliferation of cardiomyocytes from iPSCs characterized by higher ratio EdU positive cells in differentiated cardiomyocytes. We next observed that IGFBP4 inhibited β-catenin expression in cytosol of EBs at late stage during differentiation of iPSCs. Knockdown of β-catenin using a siRNA technique promoted the proliferation of differentiated cardiomyocytes and enhanced cardiomyocytes induction of iPSCs, suggesting that the effect of IGFBP4 on cardiomyocytes differentiation of iPSCs has relationship with β-catenin signaling pathway. In conclusion, IGFBP4 promotes cardiogenesis of iPSCs by enhancing the proliferation of differentiated cardiomyocytes through inhibiting β-catenin signaling.
Sigma-Aldrich
Monoclonal Anti-GJA1 antibody produced in mouse, clone 3E5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland