Merck
  • Extraction optimization of Loratadine by supramolecular solvent-based microextraction and its determination using HPLC.
Extraction optimization of Loratadine by supramolecular solvent-based microextraction and its determination using HPLC.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-13)
Moazameh Peyrovi, Mohammadreza Hadjmohammadi
ABSTRACT
Optimization of supramolecular solvent-based microextraction (SSME) of Loratadine and its determination with high-performance liquid chromatography (HPLC) with ultra violet (UV) detection were investigated. A factorial design (FD) and a central composite face-centered (CCF) were applied to evaluate the SSME procedure. The effect of four parameters on extraction efficiency was investigated. The factors studied were decanoic acid amount, percentage of tetrahydrofuran (THF) (v/v), pH and extraction time. According to half factorial design results, the effective parameters were decanoic acid amount, THF percentage (v/v) and pH. Then, a CCF was applied to obtain optimal condition. The optimized conditions were obtained at 110mg of decanoic acid, 10% of THF and pH=3. The limits of detection were in the range of 0.3-0.4ng/ml. Linearity of the method was determined to be in the range of 1.0-400.0ng/ml for distilled water and 1.3-400.0ng/ml for plasma. The extraction recovery was >92%. RSD for intra and inter day (n=5) of extraction of Loratadine were 3.1% and 6.2%, respectively. The developed method was successfully applied for the determination of Loratadine in distilled water and plasma samples.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Supelco
Tetrahydrofuran, analytical standard
Supelco
Decanoic acid, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Decanoic acid, ≥98.0%
USP
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%