Merck
  • Proteomic study reveals a co-occurrence of gallic acid-induced apoptosis and glycolysis in B16F10 melanoma cells.
Proteomic study reveals a co-occurrence of gallic acid-induced apoptosis and glycolysis in B16F10 melanoma cells.
Journal of agricultural and food chemistry (2014-11-15)
Cheng Liu, Jen-Jie Lin, Zih-Yan Yang, Chi-Chu Tsai, Jue-Liang Hsu, Yu-Jen Wu
ABSTRACT
Gallic acid (GA) has long been associated with a wide range of biological activities. In this study, its antitumor effect against B16F10 melanoma cells was demonstrated by MTT assay, cell migration assay, wound-healing assay, and flow cytometric analysis. GA with a concentration >200 μM shows apoptotic activity toward B16F10 cells. According to Western blotting data, overexpressions of cleaved forms of caspase-9, caspase-3, and PARP-1 and pro-apoptotic Bax and Bad, accompanied by underexpressed anti-apoptotic Bcl-2 and Bcl-xL indicate that GA induces B16F10 cell apoptosis via mitochondrial pathway. The 2-DE based comparative proteomics was further employed in B16F10 cells with and without GA treatment for a large-scale protein expression profiling. A total of 41 differential protein spots were quantified, and their identities were characterized using LC-MS/MS analysis and database matching. In addition to some regulated proteins that were associated with apoptosis, interestingly, some identified proteins involved in glycolysis such as glucokinase, α-enolase, aldolase, pyruvate kinase, and GAPDH were simultaneously up-regulated, which reveals that the GA-induced cellular apoptosis in B16 melanoma cells is associated with metabolic glycolysis.
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, Molecular Biology, ≥99.5% (RT)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)