Merck
  • C/EBP homologous protein inhibits tissue repair in response to gut injury and is inversely regulated with chronic inflammation.
C/EBP homologous protein inhibits tissue repair in response to gut injury and is inversely regulated with chronic inflammation.
Mucosal immunology (2014-05-23)
N Waldschmitt, E Berger, E Rath, R B Sartor, B Weigmann, M Heikenwalder, M Gerhard, K-P Janssen, D Haller
ABSTRACT
Loss of intestinal epithelial cell (IEC) homeostasis and apoptosis negatively affect intestinal barrier function. Uncontrolled activation of the unfolded protein response (UPR) in IEC contributes to an impaired barrier and is implicated in the pathogenesis of inflammatory bowel diseases. However, the contribution of the UPR target gene C/EBP homologous protein (CHOP), an apoptosis-associated transcription factor, to inflammation-related disease susceptibility remains unclear. Consistent with observations in patients with ulcerative colitis, we show that despite UPR activation in the epithelium, CHOP expression was reduced in mouse models of T-cell-mediated and bacteria-driven colitis. To elucidate the molecular mechanisms of IEC-specific CHOP expression, we generated a conditional transgenic mouse model (Chop(IEC Tg/Tg)). Chop overexpression increased the susceptibility toward dextran sodium sulfate (DSS)-induced intestinal inflammation and mucosal tissue injury. Furthermore, a delayed recovery from DSS-induced colitis and impaired closure of mechanically induced mucosal wounds was observed. Interestingly, these findings seemed to be independent of CHOP-mediated apoptosis. In vitro and in vivo cell cycle analyses rather indicated a role for CHOP in epithelial cell proliferation. In conclusion, these data show that IEC-specific overexpression impairs epithelial cell proliferation and mucosal tissue regeneration, suggesting an important role for CHOP beyond mediating apoptosis.
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-GRP78/BiP (ET-21) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Methylene blue, certified by the BSC