Merck
  • Chitosan/glucose 1-phosphate as new stable in situ forming depot system for controlled drug delivery.
Chitosan/glucose 1-phosphate as new stable in situ forming depot system for controlled drug delivery.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-05-27)
Stephanie Supper, Nicolas Anton, Julie Boisclair, Nina Seidel, Marc Riemenschnitter, Catherine Curdy, Thierry Vandamme
ABSTRACT
Chitosan (CS)-based thermosensitive solutions that turn into semi-solid hydrogels upon injection at body temperature have increasingly drawn attention over the last decades as an attractive new type of in situ forming depot (ISFD) drug delivery system. Despite the great potential of the standard CS/β-glycerophosphate (β-GP) thermogelling solutions, their lack of stability over time at room temperature as well as at refrigerated conditions renders them unsuitable as ready-to-use drug product. In the present study, we investigated Glucose-1-Phosphate (G1-P) as an alternative gelling agent for improving the stability of CS-based ISFD solutions. The in vitro release performance of CS/G1-P formulations was assessed using several model compounds. Furthermore, the local tolerance of subcutaneously implanted CS/G1-P hydrogels was investigated by histological examination over three weeks. The thermogelling potential of CS/G1-P solutions, determined by rheology, is dependent on the polymer molecular weight (Mw) and concentration as well as on the G1-P concentration. Differential scanning calorimetry (DSC) measurements confirmed that sol/gel transition takes place at around body temperature and is not fully thermo-reversible. The long term storage stability was evaluated through the appearance, pH, viscosity and gelation time at 37°C of the solution. The results emphasized an enhanced stability of the CS/G1-P system compared to the standard CS/β-GP. CS solution with 0.40 mmol/g G1-P is stable for at least 9 months at 2-8°C, versus less than 1 month when using β-GP as gelling agent. Furthermore, the solution is easy to inject, as evidenced from injectability evaluation using 23-30 G needles. In vitro release experiments showed a sustained release over days to weeks for hydrophilic model compounds, demonstrating thereby that CS/G1-P may be suitable for the prolonged delivery of drugs. The inflammatory reaction observed in the tissue surrounding the hydrogel in rats was a typical foreign body reaction, similar to the one observed for CS/β-GP hydrogels. These features confirm the potential of CS/G1-P solutions as an injectable ready-to-use in situ forming hydrogel.
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y disodium salt, certified by the BSC
Sigma-Aldrich
Eosin Y disodium salt, ≥85 % by HPLC
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture