Merck
  • Binge alcohol consumption aggravates oxidative stress and promotes pathogenesis of NASH from obesity-induced simple steatosis.
Binge alcohol consumption aggravates oxidative stress and promotes pathogenesis of NASH from obesity-induced simple steatosis.
Molecular medicine (Cambridge, Mass.) (2014-09-03)
Takahiro Minato, Mikihiro Tsutsumi, Mutsumi Tsuchishima, Nobuhiko Hayashi, Takashi Saito, Yasuhiro Matsue, Nobuyuki Toshikuni, Tomiyasu Arisawa, Joseph George
ABSTRACT
The pathogenesis of nonalcoholic steatohepatitis (NASH) is a two-stage process in which steatosis is the "first hit" and an unknown "second hit." We hypothesized that "a binge" could be a "second hit" to develop NASH from obesity-induced simple steatosis. Thirty-week-old male Otsuka Long-Evans Tokushima fatty (OLETF) rats were administered 10 mL of 10% ethanol orally for 5, 3, 2, and 1 d/wk for 3 consecutive weeks. As control, male Otsuka Long-Evans Tokushima (OLET) rats were administered the same amount of alcohol. Various biochemical parameters of obesity, steatosis and NASH were monitored in serum and liver specimens in untreated and ethanol-treated rats. The liver sections were evaluated for histopathological alterations of NASH and stained for cytochrome P-4502E1 (CYP2E1) and 4-hydroxy-nonenal (4-HNE). Simple steatosis, hyperinsulinemia, hyperglycemia, insulin resistance, hypertriglycemia and marked increases in hepatic CYP2E1 and 4-HNE were present in 30-wk-old untreated OLETF rats. Massive steatohepatitis with hepatocyte ballooning was observed in the livers of all OLETF rats treated with ethanol. Serum and hepatic triglyceride levels as well as tumor necrosis factor (TNF)-α mRNA were markedly increased in all ethanol-treated OLETF rats. Staining for CYP2E1 and 4-NHE demonstrated marked increases in the hepatic tissue of all the groups of OLETF rats treated with ethanol compared with OLET rats. Our data demonstrated that "a binge" serves as a "second hit" for development of NASH from obesity-induced simple steatosis through aggravation of oxidative stress. The enhanced levels of CYP2E1 and increased oxidative stress in obesity play a significant role in this process.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol Standard Solution, 2.5 mg/ml equivalent triolein concentration
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Triglyceride Reagent, used for quantitative enzymatic determination of triglyerides
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Supelco
Glycerol, analytical standard