Merck
  • Lipids, tocopherols, and carotenoids in leaves of amaranth and quinoa cultivars and a new approach to overall evaluation of nutritional quality traits.
Lipids, tocopherols, and carotenoids in leaves of amaranth and quinoa cultivars and a new approach to overall evaluation of nutritional quality traits.
Journal of agricultural and food chemistry (2014-12-04)
Yao Tang, Xihong Li, Peter X Chen, Bing Zhang, Marta Hernandez, Hua Zhang, Massimo F Marcone, Ronghua Liu, Rong Tsao
ABSTRACT
Composition of lipophilic phytochemicals including fatty acids, tocopherols, and carotenoids in leaves of 6 quinoa and 14 amaranth cultivars was analyzed. The oil yields in quinoa and amaranth leaves were only 2.72-4.18%, which contained mainly essential fatty acids and had a highly favorable ω-3/ω-6 ratio (2.28-3.89). Pro-vitamin A carotenoids, mainly α- and β-carotenes, and xanthophylls, predominantly lutein and violaxanthin, were found in all samples. The primary tocopherol isomers present in both quinoa and amaranth leaves were α- and β-tocopherols. Added to the discussion on the lipophilic nutrients was the normalization of ω-3/ω-6 ratio, α-tocopherol equivalents, and carotenoids, in an attempt to establish a novel system for evaluation of the overall quality attributes of lipophilic nutrients (NQ value). The NQ value, but not the individual components, was highly correlated with all the antioxidant activities, supporting the ranking order of the potential nutritional quality of quinoa and amaranth leaves based on this new method.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionamidine) dihydrochloride, powder or granules, 97%
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
β-Carotene, synthetic, ≥95% (HPLC), crystalline
Sigma-Aldrich
β-Carotene, synthetic, ≥93% (UV), powder
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%
Sigma-Aldrich
Xanthophyll, from marigold
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Ascorbic acid, analytical standard
Supelco
β-Carotene, Pharmaceutical Secondary Standard; Certified Reference Material
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
LDN-57444, ≥98% (HPLC)
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Supelco
Zeaxanthin, analytical standard