Merck
  • Development of sample clean up methods for the analysis of Mycobacterium tuberculosis methyl mycocerosate biomarkers in sputum extracts by gas chromatography-mass spectrometry.
Development of sample clean up methods for the analysis of Mycobacterium tuberculosis methyl mycocerosate biomarkers in sputum extracts by gas chromatography-mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-03)
Simona C Nicoara, Nicholas W Turner, David E Minnikin, Oona Y-C Lee, Denise M O'Sullivan, Ruth McNerney, Reggie Mutetwa, Liz E Corbett, Geraint H Morgan
ABSTRACT
A proof of principle gas chromatography-mass spectrometry method is presented, in combination with clean up assays, aiming to improve the analysis of methyl mycocerosate tuberculosis biomarkers from sputum. Methyl mycocerosates are generated from the transesterification of phthiocerol dimycocerosates (PDIMs), extracted in petroleum ether from sputum of tuberculosis suspect patients. When a high matrix background is present in the sputum extracts, the identification of the chromatographic peaks corresponding to the methyl derivatives of PDIMs analytes may be hindered by the closely eluting methyl ether of cholesterol, usually an abundant matrix constituent frequently present in sputum samples. The purification procedures involving solid phase extraction (SPE) based methods with both commercial Isolute-Florisil cartridges, and purpose designed molecularly imprinted polymeric materials (MIPs), resulted in cleaner chromatograms, while the mycocerosates are still present. The clean-up performed on solutions of PDIMs and cholesterol standards in petroleum ether show that, depending on the solvent mix and on the type of SPE used, the recovery of PDIMs is between 64 and 70%, whilst most of the cholesterol is removed from the system. When applied to petroleum ether extracts from representative sputum samples, the clean-up procedures resulted in recoveries of 36-68% for PDIMs, allowing some superior detection of the target analytes.
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile) solution, 0.2 M in toluene
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), purum, ≥98.0% (GC)
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Trimethylphenylammonium hydroxide solution, ~0.5 M (CH3)3N(OH)C6H5 in methanol, derivatization grade (GC derivatization), LiChropur
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material