Merck
  • Oxidative stress protection by exogenous delivery of rhHsp70 chaperone to the retinal pigment epithelium (RPE), a possible therapeutic strategy against RPE degeneration.
Oxidative stress protection by exogenous delivery of rhHsp70 chaperone to the retinal pigment epithelium (RPE), a possible therapeutic strategy against RPE degeneration.
Pharmaceutical research (2014-07-18)
Astrid Subrizi, Elisa Toropainen, Eva Ramsay, Anu J Airaksinen, Kai Kaarniranta, Arto Urtti
ABSTRACT
To measure the cytoprotective effects of rhHsp70 against oxidative stress and study its cellular uptake, intracellular and intraocular distribution in the retinal pigment epithelium. Human retinal pigment epithelial cells (ARPE-19) were pre-treated with rhHsp70 for 24 h, 48 h, and 72 h before being exposed to 1.25 mM hydrogen peroxide. Non-treated cells served as control. We analysed interleukin 6 secretion, cell viability, and cytolysis. Uptake and intracellular distribution of fluorescently labelled rhHsp70 were investigated with flow cytometry and confocal microscopy, respectively. Ocular distribution of radioactively labelled rhHsp70 was followed ex vivo in porcine eyes by micro SPECT/CT. After exposure to hydrogen peroxide, IL-6 secretion decreased by 35-39% when ARPE-19 cells were pre-treated with rhHsp70. Cell viability increased by 17-32%, and cell lysis, measured by the release of lactate dehydrogenase, decreased by 6-43%. ARPE-19 cells endocytosed rhHsp70 added to the culture medium and the protein was localized in late endosomes and lysosomes. Following intravitreal injection into isolated porcine eyes, we found 20% rhHsp70 in the RPE. Recombinant hHsp70 protein offers protection against oxidative stress. RPE cells take up the exogenously delivered rhHsp70 and localize it in late endosomes and lysosomes. This work provides the basis for a therapeutic strategy to target aggregate-associated neurodegeneration in AMD.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets