Merck
  • Effects of methylglyoxal on human cardiac fibroblast: roles of transient receptor potential ankyrin 1 (TRPA1) channels.
Effects of methylglyoxal on human cardiac fibroblast: roles of transient receptor potential ankyrin 1 (TRPA1) channels.
American journal of physiology. Heart and circulatory physiology (2014-08-31)
Gaku Oguri, Toshiaki Nakajima, Yumiko Yamamoto, Nami Takano, Tomofumi Tanaka, Hironobu Kikuchi, Toshihiro Morita, Fumitaka Nakamura, Tatsuya Yamasoba, Issei Komuro
ABSTRACT
Cardiac fibroblasts contribute to the pathogenesis of cardiac remodeling. Methylglyoxal (MG) is an endogenous carbonyl compound produced under hyperglycemic conditions, which may play a role in the development of pathophysiological conditions including diabetic cardiomyopathy. However, the mechanism by which this occurs and the molecular targets of MG are unclear. We investigated the effects of MG on Ca(2+) signals, its underlying mechanism, and cell cycle progression/cell differentiation in human cardiac fibroblasts. The conventional and quantitative real-time RT-PCR, Western blot, immunocytochemical analysis, and intracellular Ca(2+) concentration [Ca(2+)]i measurement were applied. Cell cycle progression was assessed using the fluorescence activated cell sorting. MG induced Ca(2+) entry concentration dependently. Ruthenium red (RR), a general cation channel blocker, and HC030031, a selective transient receptor potential ankyrin 1 (TRPA1) antagonist, inhibited MG-induced Ca(2+) entry. Treatment with aminoguanidine, a MG scavenger, also inhibited it. Allyl isothiocyanate, a selective TRPA1 agonist, increased Ca(2+) entry. The use of small interfering RNA to knock down TRPA1 reduced the MG-induced Ca(2+) entry as well as TRPA1 mRNA expression. The quantitative real-time RT-PCR analysis showed the prominent existence of TRPA1 mRNA. Expression of TRPA1 protein was confirmed by Western blotting and immunocytochemical analyses. MG promoted cell cycle progression from G0/G1 to S/G2/M, which was suppressed by HC030031 or RR. MG also enhanced α-smooth muscle actin expression. The present results suggest that methylglyoxal activates TRPA1 and promotes cell cycle progression and differentiation in human cardiac fibroblasts. MG might participate the development of pathophysiological conditions including diabetic cardiomyopathy via activation of TRPA1.
Sigma-Aldrich
HC-030031, ≥98% (HPLC), powder
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Allyl isothiocyanate, 95%
Sigma-Aldrich
Allyl isothiocyanate, ≥95%, FCC
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Ruthenium Red, Technical grade
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Supelco
Allyl isothiocyanate, PESTANAL®, analytical standard
Supelco
DL-Dithiothreitol solution, 1 M in H2O