Merck
  • pH-sensitive nanoparticles for colonic delivery of curcumin in inflammatory bowel disease.
pH-sensitive nanoparticles for colonic delivery of curcumin in inflammatory bowel disease.
International journal of pharmaceutics (2014-07-12)
Ana Beloqui, Régis Coco, Patrick B Memvanga, Bernard Ucakar, Anne des Rieux, Véronique Préat
ABSTRACT
Nano-scaled particles have been found to preferentially accumulate in inflamed regions. Local delivery of anti-inflammatory drugs loaded in nanoparticles to the inflamed colonic site is of great interest for inflammatory bowel disease (IBD) treatment. Curcumin (CC) is an anti-inflammatory local agent, which presents poor ADME properties. Hence, we evaluated, both in vitro and in vivo, the local delivery of CC using pH-sensitive polymeric nanoparticles (NPs) combining both poly(lactide-co-glycolide) acid (PLGA) and a polymethacrylate polymer (Eudragit(®) S100). CC-NPs significantly enhanced CC permeation across Caco-2 cell monolayers when compared to CC in suspension. CC-NPs significantly reduced TNF-α secretion by LPS-activated macrophages (J774 cells). In vivo, CC-NPs significantly decreased neutrophil infiltration and TNF-α secretion while maintaining the colonic structure similar to the control group in a murine DSS-induced colitis model. Our results support the use of nanoparticles made of PLGA and Eudragit(®) S100 combination for CC delivery in IBD treatment.
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard