Merck
  • On-line coupling of solid-phase extraction to gas chromatography-mass spectrometry to determine musk fragrances in wastewater.
On-line coupling of solid-phase extraction to gas chromatography-mass spectrometry to determine musk fragrances in wastewater.
Journal of chromatography. A (2014-09-23)
Laura Vallecillos, Francesc Borrull, Eva Pocurull
ABSTRACT
An on-line coupling solid-phase extraction (SPE) has been developed for the first time to preconcentrate trace amounts of 17 musk fragrances extensively used in personal care products (6 polycyclic musks, 3 nitro musks, 7 macrocyclic musks and 1 polycyclic musk degradation product) from wastewater samples, prior to analysis by gas chromatography and mass spectrometry through an on-column interface. A 10 mm × 2 mm I.D. precolumn packed with Oasis HLB (60 μm) or C18 (60 μm) was compared for the optimization of the solid-phase extraction process. The parameters affecting the transfer of the analytes from the precolumn to the GC system (e.g. flow-rate, temperature and solvent vapor exit time) as well as SPE parameters (e.g. sample flow, sample volume, elution solvent, etc.) were optimized. An organic modifier such as methanol was added to the sample before the extraction process to avoid adsorption problems. The use of the MS detector under selected ion monitoring acquisition enabled the analytes to be quantified at low ng L(-1) levels, preconcentrating only 10 mL of sample, and the limits of detection were between 1 and 30 ng L(-1). The method was applied for the determination of musk fragrances in wastewater samples from three urban wastewater treatment plants (WWTPs). The analysis of influent urban wastewater revealed that galaxolide, tonalide and ambrettolide were the most abundant musk compounds with concentrations ranging between 18 ng L(-1) and 45,091 ng L(-1), 852 ng L(-1) and 49,904 ng L(-1) and 507 ng L(-1) and 21,528 ng L(-1) respectively. The remaining musks were present at lower concentrations and two of the macrocyclic musk studied (musk MC4 and civetone) were not detected. The analysis of effluent wastewater showed a decrease in the concentrations of all of the compounds present in influent samples, with the decrease being more significantly in the case of polycyclic and nitro musks than for macrocyclic musks. Only HHCB-lactone remained constant or increased its concentration.
Product Description
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Supelco
o-Xylene, analytical standard
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%