Merck
  • Impact of taurine supplementation on blood pressure in gestational protein-restricted offspring: Effect on the medial solitary tract nucleus cell numbers, angiotensin receptors, and renal sodium handling.
Impact of taurine supplementation on blood pressure in gestational protein-restricted offspring: Effect on the medial solitary tract nucleus cell numbers, angiotensin receptors, and renal sodium handling.
Journal of the renin-angiotensin-aldosterone system : JRAAS (2013-03-08)
José Eduardo Scabora, Marcelo Cardoso de Lima, Agnes Lopes, Ize Penhas de Lima, Flávia Fernandes Mesquita, Daniele Bráz Torres, Patrícia Aline Boer, José Antonio Rocha Gontijo
ABSTRACT
The current study considers changes of the postnatal brainstem cell number and angiotensin receptors by maternal protein restriction (LP) and LP taurine supplementation (LPT), and its impact on arterial hypertension development in adult life. The brain tissue studies were performed by immunoblotting, immunohistochemistry, and isotropic fractionator analysis. The current study shows that elevated blood pressure associated with decreased fractional urinary sodium excretion (FENa) in adult LP offspring was reverted by diet taurine supplementation. Also, that 12-day-old LP pups present a reduction of 21% of brainstem neuron counts, and, immunohistochemistry demonstrates a decreased expression of type 1 angiotensin II receptors (AT1R) in the entire medial solitary tract nuclei (nTS) of 16-week-old LP rats compared to age-matched NP and LPT offspring. Conversely, the immunostained type 2 AngII (AT2R) receptors in 16-week-old LP nTS were unchanged. The present investigation shows a decreased FENa that occurs despite unchanged creatinine clearance. It is plausible to hypothesize an association of decreased postnatal nTS cell number, AT1R/AT2R ratio and FENa with the higher blood pressure levels found in taurine-deficient progeny (LP) compared with age-matched NP and LPT offspring.
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
Glycerol, analytical standard
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material