Merck
  • The Intelence aNd pRezista Once A Day Study (INROADS): a multicentre, single-arm, open-label study of etravirine and darunavir/ritonavir as dual therapy in HIV-1-infected early treatment-experienced subjects.
The Intelence aNd pRezista Once A Day Study (INROADS): a multicentre, single-arm, open-label study of etravirine and darunavir/ritonavir as dual therapy in HIV-1-infected early treatment-experienced subjects.
HIV medicine (2015-01-15)
P J Ruane, C Brinson, M Ramgopal, R Ryan, B Coate, M Cho, T N Kakuda, D Anderson
ABSTRACT
Following antiretroviral therapy failure, patients are often treated with a three-drug regimen that includes two nucleoside/tide reverse transcriptase inhibitors [N(t)RTIs]. An alternative two-drug nucleoside-sparing regimen may decrease the pill burden and drug toxicities associated with the use of N(t)RTIs. The Intelence aNd pRezista Once A Day Study (INROADS; NCT01199939) evaluated the nucleoside-sparing regimen of etravirine 400 mg with darunavir/ritonavir 800/100 mg once-daily in HIV-1-infected treatment-experienced subjects or treatment-naïve subjects with transmitted resistance. In this exploratory phase 2b, single-arm, open-label, multicentre, 48-week study, the primary endpoint was the proportion of subjects who achieved HIV-1 RNA < 50 copies/mL at week 48 [confirmed virological response (CVR), non-virological failure (VF) censored]. Key secondary endpoints included assessments of changes from baseline to week 48 in viral load, immunological response, pharmacokinetics/pharmacodynamics, safety, tolerability, metabolic and bone markers and body fat. Forty-one of the 54 enrolled subjects completed the study. Adverse events (7%) and VF (7%) were the most common reasons for discontinuation. The week 48 CVR rate in the intent-to-treat (ITT) non-VF censored population was 89% (primary endpoint). Seven subjects experienced VF. Common adverse events were diarrhoea (15%), rash (15%) and upper respiratory tract infection (11%). Mild/moderate lipid elevations, minimal changes in limb fat distribution and bone mineral density and no clinically relevant changes in glucose metabolism were observed. Etravirine 400 mg and darunavir/ritonavir 800/100 mg as a two-drug once-daily regimen in treatment-experienced subjects or treatment-naïve subjects with transmitted resistance was virologically efficacious and well tolerated.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications