Merck
Lumazine peptides from the marine-derived fungus Aspergillus terreus.
Marine drugs (2015-03-17)
Minjung You, Lijuan Liao, Soo Hyun Hong, Wanki Park, Dah In Kwon, Jeeyeon Lee, Minsoo Noh, Dong-Chan Oh, Ki-Bong Oh, Jongheon Shin
ABSTRACT
Terrelumamides A (1) and B (2), two new lumazine-containing peptides, were isolated from the culture broth of the marine-derived fungus Aspergillus terreus. From the results of combined spectroscopic and chemical analyses, the structures of these compounds were determined to be linear assemblies of 1-methyllumazine-6-carboxylic acid, an amino acid residue and anthranilic acid methyl ester connected by peptide bonds. These new compounds exhibited pharmacological activity by improving insulin sensitivity, which was evaluated in an adipogenesis model using human bone marrow mesenchymal stem cells. In addition, the compounds exhibited fluorescence changes upon binding to DNA, demonstrating their potential applications to DNA sequence recognition.
Product Description
Supelco
3-Nitrobenzyl alcohol, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
3-Nitrobenzyl alcohol, 98%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS