Merck
  • Modulation of cell metabolic pathways and oxidative stress signaling contribute to acquired melphalan resistance in multiple myeloma cells.
Modulation of cell metabolic pathways and oxidative stress signaling contribute to acquired melphalan resistance in multiple myeloma cells.
PloS one (2015-03-15)
Kamila Anna Zub, Mirta Mittelstedt Leal de Sousa, Antonio Sarno, Animesh Sharma, Aida Demirovic, Shalini Rao, Clifford Young, Per Arne Aas, Ida Ericsson, Anders Sundan, Ole Nørregaard Jensen, Geir Slupphaug
ABSTRACT
Alkylating agents are widely used chemotherapeutics in the treatment of many cancers, including leukemia, lymphoma, multiple myeloma, sarcoma, lung, breast and ovarian cancer. Melphalan is the most commonly used chemotherapeutic agent against multiple myeloma. However, despite a 70-80% initial response rate, virtually all patients eventually relapse due to the emergence of drug-resistant tumour cells. By using global proteomic and transcriptomic profiling on melphalan sensitive and resistant RPMI8226 cell lines followed by functional assays, we discovered changes in cellular processes and pathways not previously associated with melphalan resistance in multiple myeloma cells, including a metabolic switch conforming to the Warburg effect (aerobic glycolysis), and an elevated oxidative stress response mediated by VEGF/IL8-signaling. In addition, up-regulated aldo-keto reductase levels of the AKR1C family involved in prostaglandin synthesis contribute to the resistant phenotype. Finally, selected metabolic and oxidative stress response enzymes were targeted by inhibitors, several of which displayed a selective cytotoxicity against the melphalan-resistant cells and should be further explored to elucidate their potential to overcome melphalan resistance.
Product Description
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dichloroacetic acid, ReagentPlus®, ≥99%
Supelco
Ibuprofen
Sigma-Aldrich
DCA Deblock (0.36M dichloroacetic acid in toluene)
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Supelco
Flufenamic acid, analytical standard, for drug analysis
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), BioXtra
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Dichloroacetic acid, PESTANAL®, analytical standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
FLLL31, ≥98% (HPLC)
Sigma-Aldrich
Luperox® DI, tert-Butyl peroxide, 98%
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Methylglyoxal solution, ~40% in H2O