Merck
  • Hydrocellular foam dressings promote wound healing associated with decrease in inflammation in rat periwound skin and granulation tissue, compared with hydrocolloid dressings.
Hydrocellular foam dressings promote wound healing associated with decrease in inflammation in rat periwound skin and granulation tissue, compared with hydrocolloid dressings.
Bioscience, biotechnology, and biochemistry (2014-11-25)
Takumi Yamane, Gojiro Nakagami, Sawako Yoshino, Mari Shimura, Aya Kitamura, Kazuo Kobayashi-Hattori, Yuichi Oishi, Yoshimi Nishijima, Takeo Minematsu, Hiromi Sanada
ABSTRACT
The effects of modern dressings on inflammation, which represent the earliest phase of wound healing, are poorly understood. We investigated the effects of modern hydrocellular foam dressings (HCFs) on wound healing and on the gene expression levels of the inflammatory markers--interleukin (IL)-1β, IL-6, and IL-10--in rat periwound skin and granulation tissue by quantitative reverse transcription-polymerase chain reaction. HCF absorbed significantly higher volume of water than hydrocolloid dressing (HCD) and increased the contraction of wounds. Polymorphonuclear neutrophils were massively infiltrated to the wound edge and boarded between granulation and dermis in the HCD group. IL-1β, IL-6, and IL-10 mRNA levels were significantly decreased in the periwound skin around the wounds and granulation tissue covered with HCF. These findings suggest that HCF may promote wound healing along with decrease in inflammation by reducing gene expression levels of IL-1β, IL-6, and IL-10.
Product Description
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard