Merck
  • Osteoblastic and osteoclastic differentiation of human mesenchymal stem cells and monocytes in a miniaturized three-dimensional culture with mineral granules.
Osteoblastic and osteoclastic differentiation of human mesenchymal stem cells and monocytes in a miniaturized three-dimensional culture with mineral granules.
Acta biomaterialia (2014-09-10)
Anne Laure Gamblin, Audrey Renaud, Céline Charrier, Philippe Hulin, Guy Louarn, Dominique Heymann, Valérie Trichet, Pierre Layrolle
ABSTRACT
The pathologies of the skeleton have a significant socioeconomic impact on our population. Although therapies have improved the treatment of osteosarcoma and osteoporosis, their efficacy still remains limited. In this context, we developed a miniaturized 3-D culture model of bone cells on calcium phosphate ceramics. Human bone marrow mesenchymal stem cells (MSCs) were three-dimensionally cultured on particles of biphasic calcium phosphate (BCP, 125-200μm) in osteogenic media. The MSCs seeded on the BCP particles adhered and proliferated, producing abundant collagenous extracellular matrix (ECM). Light and confocal laser scanning microscopy showed that the MSCs created bridges between the BCP particles and formed a 3-D structure. Energy dispersive X-ray analysis in a scanning electron microscope confirmed the mineralization of the collagen matrix. The 96-well sized bone constructs were tested by immunohistology and transcription analysis, proving cell differentiation. Both techniques corroborated the osteoblastic differentiation with high production of bone sialoprotein and osteocalcin. Peripheral blood CD14-positive monocytes (MOs) were pre-differentiated into osteoclasts prior to seeding on the 3-D constructs. Multinucleated and tartrate-resistant acid phosphatase-positive cells were also identified at the surface of the 3-D constructs after 90days of culture. In addition, cell viability within these constructs was measured by flow cytometry. In summary, we have developed a miniaturized 3-D culture of bone cell precursors with osteoblasts and osteoclasts. This 3-D culture may make it possible to test the effects of new drugs for bone healing, osteoporosis and osteosarcomas, in more appropriate cell-cell and cell-matrix interactions than conventional 2-D cultures.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Sulfobromophthalein disodium salt hydrate, used to study hepatocyte transport functions