Merck
  • Nanoplasmonic biosensor: detection and amplification of dual bio-signatures of circulating tumor DNA.
Nanoplasmonic biosensor: detection and amplification of dual bio-signatures of circulating tumor DNA.
Biosensors & bioelectronics (2014-09-16)
Anh H Nguyen, Sang Jun Sim
ABSTRACT
Circulating tumor DNA (ctDNA) bearing tumor-specific mutation and methylation are promising biomarkers for noninvasive cancer assessment. However, existing methods for ctDNA detection are restricted to genetic mutations. Recently, nanoplasmonics has emerged as a platform for one-step dual detection with high sensitivity and specificity. Here we present a strategy for ultrasensitive detection of tumor-specific mutations (E542K and E545K) and methylation of ctDNA of PIK3CA gene based on localized surface plasmon resonance (LSPR) and the coupling plasmon mode of gold nanoparticles (AuNPs). Peptide nucleic acids (PNA) is used as a probe to capture and enrich the 69-bp PIK3CA ctDNA. The exposure of PNA-probed AuNPs to 200 fM ctDNA generates LSPR-peak shift of 4.3 nm, corresponding to the primary response. Immunogold colloids are exploited as methylation detectors and plasmon coupling based enhancement for secondary response. LSPR-peak shifted from 4.3 nm to 11.4 nm upon the immunogold colloids binding to two methylcytosines (mCpG), which is an approximately 107% increase, compared to that of the primary response. This enhancement leads to four times (~50 fM) improvement of sensitivity and because of two mCpG sites, ctDNA was detected. These results demonstrate that the sensor can simultaneously detect the hot-spot mutation and epigenetic changes on the ctDNA. Promisingly, other specific-tumor mutants and epigenetic changes can be detected at low concentration with this platform.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ≥98%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%