Merck
  • Paricalcitol, a vitamin d receptor activator, inhibits tumor formation in a murine model of uterine fibroids.
Paricalcitol, a vitamin d receptor activator, inhibits tumor formation in a murine model of uterine fibroids.
Reproductive sciences (Thousand Oaks, Calif.) (2014-06-14)
Sunil K Halder, Chakradhari Sharan, Omar Al-Hendy, Ayman Al-Hendy
ABSTRACT
We examined the antitumor and therapeutic potentials of paricalcitol, an analog of 1,25-dihydroxyvitamin D3 with lower calcemic activity, against uterine fibroids using in vitro and in vivo evaluations in appropriate uterine fibroid cells and animal models. We found that paricalcitol has potential to reduce the proliferation of the immortalized human uterine fibroid cells. For the in vivo study, we generated subcutaneous tumors by injecting the Eker rat-derived uterine leiomyoma cell line (ELT-3) rat uterine fibroid-derived cell line in athymic nude mice supplemented with estrogen pellets. These mice were administered with vehicle versus paricalcitol (300 ng/kg/d) or 1,25-dihydroxyvitamin D3 (500 ng/kg/d) for 4 consecutive weeks, and the data were analyzed. We found that while both paricalcitol and 1,25-dihydroxyvitamin D3 significantly reduced fibroid tumor size, the shrinkage was slightly higher in the paricalcitol-treated group. Together, our results suggest that paricalcitol may be a potential candidate for effective, safe, and noninvasive medical treatment option for uterine fibroids.
Product Description
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
USP
Paricalcitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, analytical standard
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®