Merck
  • Expression of Sox2 in breast cancer cells promotes the recruitment of M2 macrophages to tumor microenvironment.
Expression of Sox2 in breast cancer cells promotes the recruitment of M2 macrophages to tumor microenvironment.
Cancer letters (2014-12-03)
Wenjun Mou, Yingxi Xu, Yujie Ye, Si Chen, Xuefei Li, Kangzi Gong, Yanhua Liu, Yanan Chen, Xiru Li, Yaping Tian, Rong Xiang, Na Li
ABSTRACT
Transcriptional factor Sox2 promotes tumor metastasis; however its regulatory effect on tumor-associated macrophages (TAMs, M2 phenotype) has not been defined. This study disclosed concomitant expression of TAMs marker-CD163 with SOX2 in human breast cancer and showed that Sox2 in breast cancer cells promotes recruitment of TAMs with altered expression of multiple chemokines, including MIP-1α, ICAM-1 etc. and activation of Stat3 and NF-κB signalings. In addition, TAMs rescued the compromised lung metastasis induced by Sox2 silencing in breast cancer cells. Together, this study documented that Sox2 plays an important role in recruiting TAMs and promotes tumor metastasis in a TAMs dependent manner.
Sigma-Aldrich
N-[Tris(hydroxymethyl)methyl]acrylamide, contains ≤7% KCl, 93%
Sigma-Aldrich
Ammonium pyrrolidinedithiocarbamate, ~99%
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Millipore
Brewer thioglycollate medium, NutriSelect® Plus, powder, for bacteria, pack of 500 g, suitable for microbiology