Merck
  • Enhancing effects of salt formation on catalytic activity and enantioselectivity for asymmetric hydrogenation of isoquinolinium salts by dinuclear halide-bridged iridium complexes bearing chiral diphosphine ligands.
Enhancing effects of salt formation on catalytic activity and enantioselectivity for asymmetric hydrogenation of isoquinolinium salts by dinuclear halide-bridged iridium complexes bearing chiral diphosphine ligands.
Chemistry (Weinheim an der Bergstrasse, Germany) (2014-12-03)
Yusuke Kita, Kenta Yamaji, Kosuke Higashida, Kandula Sathaiah, Atuhiro Iimuro, Kazushi Mashima
ABSTRACT
Asymmetric hydrogenation of 1- and 3-substituted and 1,3-disubstituted isoquinolinium chlorides using triply halide-bridged dinuclear iridium complexes [{Ir(H)(diphosphine)}2 (μ-Cl)3 ]Cl has been achieved by the strategy of HCl salt formation of isoquinolines to afford the corresponding chiral 1,2,3,4-tetrahydroisoquinolines (THIQs) in high yields and with excellent enantioselectivities after simple basic work-up. The effects of salt formation have been investigated by time-course experiments, which revealed that the generation of isoquinolinium chlorides clearly prevented formation of the catalytically inactive dinuclear trihydride complex, which was readily generated in the catalytic reduction of salt-free isoquinoline substrates. Based on mechanistic investigations, including by (1) H and (31) P{(1) H} NMR studies and the isolation and characterization of several intermediates, the function of the chloride anion of the isoquinolinium chlorides has been elucidated, allowing us to propose a new outer-sphere mechanism involving coordination of the chloride anion of the substrates to an iridium dihydride species along with a hydrogen bond between the chloride ligand and the N-H proton of the substrate salt.
Sigma-Aldrich
Acetamide, sublimed, 99%
Sigma-Aldrich
Acetamide, ~99% (GC)
Sigma-Aldrich
Acetamide, ≥99.0% (GC)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer