Merck
  • Genotoxic potential and in vitro tumour-promoting potential of 2-dodecylcyclobutanone and 2-tetradecylcyclobutanone, two radiolytic products of fatty acids.
Genotoxic potential and in vitro tumour-promoting potential of 2-dodecylcyclobutanone and 2-tetradecylcyclobutanone, two radiolytic products of fatty acids.
Mutation research. Genetic toxicology and environmental mutagenesis (2014-10-26)
Kohji Yamakage, Hajime Sui, Ryo Ohta, Tomoyasu Toyoizumi, Kumiko Kawakami, Hirotaka Matsumoto, Toshitaka Takahashi, Kiyoshi Sasaki, Mayu Ikezumi, Saki Negishi, Keisuke Izumi, Setsuko Todoriki, Kondo Takashi, Masakazu Furuta
ABSTRACT
The DNA-damaging and tumour-promoting effects of two 2-alkylcyclobutanones (2-ACBs), which are found in irradiated fat-containing foods, were investigated by use of the comet assay and in an azoxymethane (AOM)-induced colon-carcinogenesis study in rats, respectively. We conducted genotoxicity tests of 2-dodecylcyclobutanone (2-dDCB) and 2-tetradecylcyclobutanone (2-tDCB) according to the test guidelines for chemicals or drugs. In addition, a cell-transformation assay with Bhas 42 cells was performed to investigate their promoting potential in vitro. The Salmonella typhimurium mutagenicity assay (Ames test), conducted with five tester strains, revealed that neither 2-dDCB nor 2-tDCB possessed mutagenic activity. Moreover, both in the in vitro chromosomal aberration test on CHL/IU cells and the in vivo bone-marrow micronucleus test where mice were given 2-dDCB and 2-tDCB (orally, up to 2000 mg/kg bw/day), we did not detect any clastogenic effects. Furthermore, DNA strand-breaks were not detected in the in vitro comet assay with CHL/IU cells, and DNA adducts derived from 2-dDCB and 2-tDCB were not detected in the colon tissues of the mice used for the micronucleus tests, in rats from a repeated dose 90-day oral toxicity test (0.03% 2-tDCB in the diet), or in rats from the AOM-induced carcinogenesis study (0.025% 2-tDCB in the diet). An in vitro tumour-promotion assay with Bhas 42 cells revealed that the number of transformed foci increased significantly following treatment of cells in the stationary phase with 2-dDCB or 2-tDCB for 10 days. Our results indicate that neither 2-dDCB nor 2-tDCB were genotoxic chemicals. However, they exhibited promoting activity, at least in vitro, when Bhas 42 cells were continuously exposed to these chemicals at toxic doses.
Sigma-Aldrich
Methyl methanesulfonate, 99%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Cyclophosphamide monohydrate, ISOPAC®
Sigma-Aldrich
Cyclophosphamide monohydrate, bulk package
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Supelco
Stearic acid, analytical standard
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
Supelco
Methyl methanesulfonate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Supelco
Cyclophosphamide monohydrate, analytical standard
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
9-Aminoacridine, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC)