Merck
  • Dopamine D1 and opioid receptor antagonist-induced reductions of fructose and saccharin intake in BALB/c and SWR inbred mice.
Dopamine D1 and opioid receptor antagonist-induced reductions of fructose and saccharin intake in BALB/c and SWR inbred mice.
Pharmacology, biochemistry, and behavior (2015-01-28)
Tamar T Kraft, Donald Huang, Elona Natanova, Melanie Lolier, Yakov Yakubov, Sam La Magna, Deena Warshaw, Anthony Sclafani, Richard J Bodnar
ABSTRACT
Sugar and fat intake in rodents are mediated in part by brain dopamine (DA) and opioid neurotransmitter systems although important strain differences exist. Thus, whereas sucrose intake of BALB/c and SWR mice was reduced by DA D1 (SCH23390: SCH) receptor antagonism, opioid (naltrexone: NTX) receptor antagonism reduced intake only in BALB/c mice. Both SCH and NTX reduced fat (Intralipid) intake in SWR, but not BALB/c mice. The present study extended this pharmacological analysis to caloric and non-caloric sweeteners by examining whether fructose (8%) or saccharin (0.2%) intakes were differentially suppressed in BALB/c and SWR mice by SCH (50-1600nmol/kg) or NTX (0.01-5mg/kg) over a 5- to 120-min time course. SCH significantly reduced fructose (200-1600nmol/kg) and saccharin (50-1600nmol/kg) intakes in both strains as did NTX (0.1-5mg/kg). Antagonist ID40 potencies were <50nmol/kg for SCH and 0.9mg/kg for NTX in inhibiting saccharin intake, and 1234nmol/kg for SCH and 5mg/kg for NTX in inhibiting fructose intake in BALB/c mice. For SWR mice, the ID40 potencies were <50nmol/kg for SCH and 0.02mg/kg for NTX in inhibiting saccharin intake, and 298nmol/kg for SCH and 2.6mg/kg for NTX in inhibiting fructose intake. Thus, saccharin intake was similarly reduced by SCH and NTX in BALB/c and SWR mice, but greater potencies of opioid (1.9-fold) and DA D1 (4-fold) receptor antagonism of fructose intake were observed in SWR relative to BALB/c mice, indicating strong strain differences.
Product Description
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
Saccharin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
Saccharin, ≥98%
USP
Fructose, United States Pharmacopeia (USP) Reference Standard
USP
Saccharin, United States Pharmacopeia (USP) Reference Standard
Fructose, European Pharmacopoeia (EP) Reference Standard
Supelco
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)