Merck
  • Third-stage nematode larvae of Contracaecum osculatum from Baltic cod (Gadus morhua) elicit eosinophilic granulomatous reactions when penetrating the stomach mucosa of pigs.
Third-stage nematode larvae of Contracaecum osculatum from Baltic cod (Gadus morhua) elicit eosinophilic granulomatous reactions when penetrating the stomach mucosa of pigs.
Parasitology research (2015-01-08)
S B Strøm, S Haarder, R Korbut, H Mejer, S M Thamsborg, P W Kania, K Buchmann
ABSTRACT
Third-stage larvae of the anisakid nematode Contracaecum osculatum were recovered from livers of Atlantic cod (Gadus morhua) caught in the Baltic Sea (June 2014) and used for experimental infection of two pigs (one male and one female). Each pig received 215 larvae by oral infection (feeding with minced cod liver containing live nematode larvae). Pigs were euthanized after 5 days, necropsied, and subjected to parasitological investigation. A total of 12 larvae were found penetrating the mucosa of the ventricle (7 in the female pig and 5 in the male pig) eliciting a granulomatous reaction at the penetration site. Four non-attached larvae were found in the female pig stomach and one in the male pig. Petechial bleeding was observed at several locations in the ventricular mucosa where larvae were located. Histological examination of the stomach mucosa revealed a massive cellular infiltration (giant cells, lymphocytes, macrophages, granulocytes, and fibroblast like cells) around the penetrating larva. Mononuclear and polymorphonuclear cells containing eosinophilic granulae were particularly prominent in the granulomas. Reactions correspond to reactions in pigs following experimental infection with the human pathogenic anisakid larvae Anisakis sp. and Pseudoterranova sp. which suggests that C. osculatum might have a zoonotic potential as well.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications