Merck
  • An efficient siRNA-mediated gene silencing in primary human monocytes, dendritic cells and macrophages.
An efficient siRNA-mediated gene silencing in primary human monocytes, dendritic cells and macrophages.
Immunology and cell biology (2014-06-04)
Anthony Troegeler, Claire Lastrucci, Carine Duval, Antoine Tanne, Céline Cougoule, Isabelle Maridonneau-Parini, Olivier Neyrolles, Geanncarlo Lugo-Villarino
ABSTRACT
Mononuclear phagocytes (MP) comprise monocytes, macrophages (MΦ) and dendritic cells (DC), including their lineage-committed progenitors, which together have an eminent role in health and disease. Lipid-based siRNA-mediated gene inactivation is an established approach to investigate gene function in MP cells. However, although there are few protocols dedicated for siRNA-mediated gene inactivation in primary human DC and MΦ, there are none available for primary human monocytes. Moreover, there is no available method to perform comparative studies of a siRNA-mediated gene silencing in primary monocytes and other MP cells. Here, we describe a protocol optimized for the lipid-based delivery of siRNA to perform gene silencing in primary human blood monocytes, which is applicable to DCs, and differs from the classical route of siRNA delivery into MΦs. Along with this protocol, we provide a comparative analysis of how monocytes, DC and MΦ are efficiently transfected with the target siRNA without affecting cell viability, resulting in strong gene knockdown efficiency, including the simultaneous inactivation of two genes. Moreover, siRNA delivery does not affect classical functions in MP such as differentiation, phagocytosis and migration, demonstrating that this protocol does not induce non-specific major alterations in these cells. As a proof-of-principle, a functional analysis of hematopoietic cell kinase (Hck) shows for the first time that this kinase regulates the protease-dependent migration mode in human monocytes. Collectively, this protocol enables efficient gene inactivation in primary MP, suggesting a wide spectrum of applications such as siRNA-based high-throughput screening, which could ultimately improve our knowledge about MP biology.
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Hck
Sigma-Aldrich
MISSION® esiRNA, targeting human HCK
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%